Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10056392
Description
TRANCE/TNFSF11/RANK L Polyclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TRANCE/TNFSF11/RANK L | |
Polyclonal | |
Unconjugated | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
The amino acid sequence used is 88% homologous in human and 94% homologous in rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
0.5 mg/mL | |
Western Blot 0.5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 30082761) | |
NP_003692 | |
TNFSF11 | |
A synthetic peptide corresponding to amino acids 161-177 of mouse CD254 (AQPFAHLTINAASIPSG) was used as immunogen for this antibody. | |
0.1 mg | |
Cancer | |
8600 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction