Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $427.50
Specifications
Antigen | TRANCE/TNFSF11/RANK L |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 30082761) |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056392T
![]() |
Novus Biologicals
NB10056392SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056392
![]() |
Novus Biologicals
NB10056392 |
0.1 mg |
Each for $427.50
|
|
|||||
Description
TRANCE/TNFSF11/RANK L Polyclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TRANCE/TNFSF11/RANK L | |
Western Blot 0.5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 30082761) | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
NP_003692 | |
8600 | |
A synthetic peptide corresponding to amino acids 161-177 of mouse CD254 (AQPFAHLTINAASIPSG) was used as immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
TNFSF11 | |
IgG | |
Protein G purified | |
The amino acid sequence used is 88% homologous in human and 94% homologous in rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title