Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRANCE/TNFSF11/RANK L Antibody (685857) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB376779 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376779 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376779 Supplier Novus Biologicals Supplier No. FAB6264Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TRANCE/TNFSF11/RANK L
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 685857
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TRANCE/TNFSF11/RANK L, Gly136-Asp317, Accession # O14788
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8600
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.