Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRANCE/TNFSF11/RANK L Antibody (685857), HRP, Novus Biologicals™
SDP

Catalog No. NB347056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347056 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347056 Supplier Novus Biologicals Supplier No. FAB6264H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRANCE/TNFSF11/RANK L Monoclonal antibody specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen TRANCE/TNFSF11/RANK L
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 685857
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TRANCE/TNFSF11/RANK L, Gly136-Asp317, Accession # O14788
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8600
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.