Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRANCE/TNFSF11/RANK L Antibody (IK22-5), Biotin, Novus Biologicals™
SDP

Catalog No. NB016640 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016640 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016640 Supplier Novus Biologicals Supplier No. NBP281123B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen TRANCE/TNFSF11/RANK L
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone IK22-5
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry
Formulation PBS
Gene Alias CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11
Gene Symbols TNFSF11
Host Species Rabbit
Immunogen Thus antibody was raised by immunising an SD rat with murine TRANCE/TNFSF11/RANK L-transfected baby hamster kidney cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8600
Test Specificity This antibody is specific for murine TRANCE/TNFSF11/RANK L.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.