Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tranilast
GREENER_CHOICE

Catalog No. p-4785688
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antiallergic; inhibits inflammatory mediator release from mast cells

Tranilast is an antiallergic via inhibition of chemical mediator release from mast cells. Shown to be an effective inhibitor of angiogenesis. Demonstrated to antagonize the effects of angiotensin II on human arteries, possibly by an interaction at the level of the AT1 receptor. Inhibits TRPV2-mediated responses; binds to Aβ40 monomers and increases Aβ40 fibrillation.

Chemical Identifiers

CAS 53902-12-8
Molecular Formula C18H17NO5

Specifications

Quantity 10 mg
Formula Weight 327.34
Purity >99%
Molecular Weight (g/mol) 327.34
Chemical Name or Material Tranilast
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.