Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Tocris Bioscience™ (±)-trans-ACPD
    
    
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                10 mg
                            
                        
                            
                                50 mg
                            
                        
                    Description
Equimolecular mixture of (1S,3R)- and (1R,3S)-ACPD. Selective agonist for metabotropic glutamate receptors; active at both group I and group II mGlu receptors (EC50 values are 2, 15, 23 and ∽800 μM at mGluR2, mGluR1, mGluR5 and mGluR4 respectively). 
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            (1S,3R)-ACPD and cis-ACPD also available.
Specifications
Specifications
| Quantity | 50 mg | 
| Formula Weight | 173.17 | 
| Purity | >99% | 
| Molecular Weight (g/mol) | 173.168 | 
| Chemical Name or Material | (±)-trans-ACPD | 
Safety and Handling
| Recommended Storage | Store at Room Temperature | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction