Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-(+)-Chrysanthemic acid, 99+%

Catalog No. ac263635000
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy

Catalog No. AC263635000

Supplier: Thermo Scientific Chemicals 263635000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 4638-92-0 | C10H16O2 | 168.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4638-92-0
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.23
MDL Number MFCD01941572
InChI Key XLOPRKKSAJMMEW-JGVFFNPUSA-N
Synonym --trans-chrysanthemic acid, 1s,3s-2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylic acid, chrysanthemic acid l-trans, l-trans-chrysanthemic acid, 1s-trans-2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylic acid, 1s-trans-2,2-dimethyl-3-2-methylprop-1-enyl cyclopropanecarboxylic acid, --trans-chrysanthemumsaeure, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methylpropenyl-, 1s-trans, 4-09-00-00169 beilstein handbook reference, chrysanthemic acid dl-trans-form
PubChem CID 33607
ChEBI CHEBI:39102
IUPAC Name (1S,3S)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylic acid
SMILES CC(=CC1C(C1(C)C)C(=O)O)C

Specifications

Melting Point 17.0°C to 21.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4768 to 1.4788
Quantity 500 mg
Merck Index 15, 2255
Specific Rotation + 14.20
Solubility Information Solubility in water: soluble.
Formula Weight 168.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material trans-(+)-Chrysanthemic acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-067-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.