Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Zeatin-riboside

Catalog No. p-7042688
Click to view available options
Quantity:
25 mg
50 mg
100 mg

CAS: 6025-53-2 | C15H21N5O5 | 351.363 g/mol

trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.

Solubility
Soluble in water, ethanol, and methanol.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6025-53-2
Molecular Formula C15H21N5O5
Molecular Weight (g/mol) 351.363
MDL Number MFCD00036809
InChI Key GOSWTRUMMSCNCW-HNNGNKQASA-N
Synonym zeatin riboside, trans-zeatin riboside, trans-zeatin-riboside, ribosylzeatin, trans-zeatinriboside, 9-ribosyl-trans-zeatin, unii-7lg4d082a9, n-4-hydroxy-3-methyl-2-butenyl adenosine, e-n-4-hydroxy-3-methyl-2-butenyl adenosine, 9-beta-d-ribosyl-trans-zeatin
PubChem CID 6440982
ChEBI CHEBI:71693
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-[[(E)-4-hydroxy-3-methylbut-2-enyl]amino]purin-9-yl]oxolane-3,4-diol
SMILES CC(=CCNC1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)O)O)CO

Specifications

Color White
Quantity 25 mg
Beilstein 5460894
Solubility Information Soluble in water,ethanol,and methanol.
Formula Weight 351.36
Physical Form Powder
Chemical Name or Material trans-Zeatin-riboside
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.