Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transcription factor E3 Antibody (IHC672), Novus Biologicals™
SDP

Catalog No. N27598701M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N27598701M 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N27598701M Supplier Novus Biologicals Supplier No. NBP2759870.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcription factor E3 Monoclonal specifically detects Transcription factor E3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Transcription factor E3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IHC672
Conjugate Unconjugated
Dilution Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
Formulation Tris Buffer, pH 7.3 - 7.7, with 1% BSA with 0.1% sodium azide
Gene Alias BHLHE33, bHLHe33transcription factor E3, Class E basic helix-loop-helix protein 33, RCCP2, TFEA, transcription factor binding to IGHM enhancer 3, transcription factor E family, member A, Transcription factor for IgH enhancer
Gene Symbols TFE3
Host Species Mouse
Immunogen Recombinant Human TFE3
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7030
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.