Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transcription factor E3 Antibody (IHC672), Novus Biologicals™

Mouse Monoclonal Antibody
$292.00
Specifications
Antigen | Transcription factor E3 |
---|---|
Clone | IHC672 |
Dilution | Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
N27598701M
![]() |
Novus Biologicals
NBP2759870.1ML |
0.1 mL |
Each of 1 for $292.00
|
|
|||||
Description
Transcription factor E3 Monoclonal specifically detects Transcription factor E3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Transcription factor E3 | |
Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
Monoclonal | |
Mouse | |
BHLHE33, bHLHe33transcription factor E3, Class E basic helix-loop-helix protein 33, RCCP2, TFEA, transcription factor binding to IGHM enhancer 3, transcription factor E family, member A, Transcription factor for IgH enhancer | |
TFE3 | |
IgG1 | |
Store at 4°C in the dark. Do not freeze. |
IHC672 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Tris Buffer, pH 7.3 - 7.7, with 1% BSA with 0.1% sodium azide | |
7030 | |
Recombinant Human TFE3 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title