Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transducin alpha Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1203504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203504 Supplier Novus Biologicals Supplier No. NB1203504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transducin alpha Polyclonal specifically detects Transducin alpha in Mouse, Porcine, Bovine, Sheep samples. It is validated for Western Blot.

Specifications

Antigen Transducin alpha
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias CSNBAD3, GBT1, GNATR, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 1, guanine nucleotide-binding protein G(t) subunit alpha-1, guanine nucleotide-binding protein G(T), alpha-1 subunit, rod-type transducin alpha subunit, Transducin alpha-1 chain, transducin, rod-specific
Gene Symbols GNAT1
Host Species Rabbit
Immunogen Synthetic Peptide: K(18) L K E D A E K D A R T V K(31) C
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2779
Test Specificity Detects Rod Transducin alpha from bovine outer rod segments and sheep retinal extract.
Target Species Mouse, Pig, Bovine, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.