Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transducin alpha Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB359505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359505 Supplier Novus Biologicals Supplier No. NB1203504MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transducin alpha Polyclonal antibody specifically detects Transducin alpha in Mouse, Porcine, Bovine, Sheep samples. It is validated for Western Blot

Specifications

Antigen Transducin alpha
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CSNBAD3, GBT1, GNATR, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 1, guanine nucleotide-binding protein G(t) subunit alpha-1, guanine nucleotide-binding protein G(T), alpha-1 subunit, rod-type transducin alpha subunit, Transducin alpha-1 chain, transducin, rod-specific
Host Species Rabbit
Immunogen Synthetic Peptide: K(18) L K E D A E K D A R T V K(31) C
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2779
Target Species Mouse, Pig, Bovine, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.