Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transducin alpha Antibody, Novus Biologicals™
SDP

Catalog No. NBP220696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220696 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220696 Supplier Novus Biologicals Supplier No. NBP220696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transducin alpha Polyclonal antibody specifically detects Transducin alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Transducin alpha
Applications Western Blot, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry-Frozen 1:100-1:1000
Formulation PBS (pH7), 20% Glycerol with 0.025% Proclin 300
Gene Accession No. P11488
Gene Alias CSNBAD3, GBT1, GNATR, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 1, guanine nucleotide-binding protein G(t) subunit alpha-1, guanine nucleotide-binding protein G(T), alpha-1 subunit, rod-type transducin alpha subunit, Transducin alpha-1 chain, transducin, rod-specific
Gene Symbols GNAT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GNAT1. The exact sequence is proprietary.
Molecular Weight of Antigen 40 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2779
Test Specificity Human. Species reactivity based on sequence identity: Canine (100%).
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.