Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Transferpette™ -8/-12 electronic Multichannel Pipette - TRADE-UP PROMO

Catalog No. p-7158866 Shop All BrandTech Scientific Products
Click to view available options
Number of Channels:
8
12
Accuracy:
±0.2μL
±0.8μL
±1.6μL
±1.8μL
Increments:
0.02μL
0.2μL

Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.

  • Choice of 8 or 12 channel models
  • Clear, legible LCD screen
  • Intuitive interface with user-friendly desing allows easy operation
  • Soft touch pipetting key and adjustable finger rest provides added comfort
  • Rechargeable battery provides over 4000 pipetting cycles between charges
  • Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
  • Easily maintained

Operating modes

  • Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
  • Mixing of samples (PIPmix Mode): program for mixing liquids
  • Reverse Pipetting (revPIP Mode): program designed for greater accuracy
  • Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
  • Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.2μL
Battery Type Rechargeable
Compatible Tips 0.1 to 20μL Nano-cap tip, 0.5 to 20μL ultra micro tip, 0.5 to 10μL tip
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Gray
Disposable Reusable
For Use With Pipette Tip (20 μL)
Grip Type Ergonomic
Material Plastic
Model Transferpette -8/-12 Electronic
Number of Channels 8
Product Line Transferpette
Programmability Step-based
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 1 to 20 μL
Display LCD
Finger Style Ergonomic
Includes Battery Pack, Universal Power Adapter with Charging Cable, Shelf Mount, Silicone O-rings, Reagent Reservoir, Grease, Operating Manual
Increments 0.02μL
Multilingual User Interface Yes
Precision 0.5% (0.1 μL)
Warranty 1 Year
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.