Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Antibody (8B9), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB359765 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359765 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359765 Supplier Novus Biologicals Supplier No. NB1107904AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transferrin Monoclonal antibody specifically detects Transferrin in Human samples. It is validated for ELISA, Immunoassay, ELISA

Specifications

Antigen Transferrin
Applications ELISA, Immunoassay, Sandwich ELISA
Classification Monoclonal
Clone 8B9
Conjugate Alexa Fluor 532
Dilution ELISA, Immunoassay, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Transferrin purified from pooled human plasma.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.