Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300340 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300340 Supplier Novus Biologicals Supplier No. NB300340

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chicken Polyclonal Antibody

Transferrin Polyclonal specifically detects Transferrin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ICC/IF (Negative).

Specifications

Antigen Transferrin
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:500, ELISA 0.1 ug/ml, ICC/IF (Negative)
Formulation PBS (pH 7.2) with 0.02% Sodium Azide
Gene Accession No. P02787
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Gene Symbols TF
Host Species Chicken
Immunogen Synthetic peptide corresponding to aa 1-698 of sertransferrin precursor protein (NP_001054.1).
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7018
Test Specificity Transferrin
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.