Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Antibody (HTF-14), Biotin, Novus Biologicals™
SDP

Catalog No. NB359288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359288 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359288 Supplier Novus Biologicals Supplier No. NB500418B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transferrin Monoclonal antibody specifically detects Transferrin in Human, Porcine, Rabbit, Bovine (Negative), Canine (Negative), Equine (Negative), Sheep (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Neutralization

Specifications

Antigen Transferrin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone HTF-14
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Radioimmunoassay, Block/Neutralize
Formulation PBS
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Host Species Mouse
Immunogen Purified porcine transferrin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Human, Pig, Rabbit, Bovine (Negative), Canine (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.