Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Antibody, Novus Biologicals™
SDP

Catalog No. NBP19747205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19747205 0.05 mg
NB018776 0.025 mg
NBP19747202 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP19747205 Supplier Novus Biologicals Supplier No. NBP1974720.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transferrin Polyclonal specifically detects Transferrin in Human, Mouse, Rat, Canine, Guinea Pig samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Transferrin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunoprecipitation 1:10-1:500
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. P02787
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Gene Symbols TF
Host Species Rabbit
Immunogen Human Transferrin.
Molecular Weight of Antigen 77 kDa
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Human, Mouse, Rat, Canine, Guinea Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.