Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Antibody, Novus Biologicals™
SDP

Catalog No. NB188092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188092 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB188092 Supplier Novus Biologicals Supplier No. NBP314752
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Transferrin Polyclonal antibody specifically detects Transferrin in Equine samples. It is validated for Immunoelectrophoresis

Specifications

Antigen Transferrin
Applications Immunoelectrophoresis
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum with 0.09% Sodium Azide
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Host Species Sheep
Immunogen no immunogen data
Quantity 2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7018
Target Species Equine
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.