Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Antibody (TW4.20), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB015916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015916 Supplier Novus Biologicals Supplier No. NBP281094AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Transferrin Monoclonal specifically detects Transferrin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Transferrin
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TW4.20
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Gene Symbols TF
Host Species Rabbit
Immunogen Iron-free purified human transferrin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7018
Test Specificity Binds to the N-terminal domain of human transferrin.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.