Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Transferrin Receptor Monoclonal Antibody (OX-26)
Mouse Monoclonal Antibody
$536.00 - $771.00
Specifications
Antigen | Transferrin Receptor |
---|---|
Clone | OX-26 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MA170033
|
Invitrogen™
MA170033 |
250 μg |
Each for $536.00
|
|
|||||
PIMA517555
|
Invitrogen™
MA517555 |
500 μg |
Each for $771.00
|
|
|||||
Description
Transferrin Receptor Monoclonal Antibody for Western Blot, IHC (F), Flow, IP
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.Specifications
Transferrin Receptor | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.02% sodium azide | |
Q99376 | |
64678 | |
PHA activated lymph node cells of a PVG rat. | |
Antibody |
OX-26 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2a | |
Rat | |
TFRC | |
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
TFRC | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title