Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transgelin/TAGLN/SM22 alpha Antibody (OTI8C8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27484A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27484A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27484A70 Supplier Novus Biologicals Supplier No. NBP274584AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transgelin/TAGLN/SM22 alpha Monoclonal specifically detects Transgelin/TAGLN/SM22 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Transgelin/TAGLN/SM22 alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI8C8
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp686P11128, Protein WS3-10, SM2222 kDa actin-binding protein, SMCCDKFZp686B01212, Smooth muscle protein 22-alpha, TAGLN1, transgelin, transgelin variant 2, WS3-10SM22-alpha
Gene Symbols TAGLN
Host Species Mouse
Immunogen Full length recombinant protein of human Transgelin/TAGLN/SM22 alpha (NP_003177) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6876.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.