Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transgelin/TAGLN/SM22 alpha Antibody (SPM606), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP247689A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP247689A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP247689A94 Supplier Novus Biologicals Supplier No. NBP247689AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transgelin/TAGLN/SM22 alpha Monoclonal specifically detects Transgelin/TAGLN/SM22 alpha in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen Transgelin/TAGLN/SM22 alpha
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone SPM606
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp686P11128, Protein WS3-10, SM2222 kDa actin-binding protein, SMCCDKFZp686B01212, Smooth muscle protein 22-alpha, TAGLN1, transgelin, transgelin variant 2, WS3-10SM22-alpha
Gene Symbols TAGLN
Host Species Mouse
Immunogen Recombinant full-length human Transgelin/TAGLN/SM22 alpha protein. (Uniprot: Q01995)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6876
Test Specificity This monoclonal antibody recognizes a 22kDa protein, identified as Transgelin, also designated SM22-alpha. It may cross-react with SM22-beta. Transgelin is expressed abundantly in smooth muscle cells. The human transgelin gene encodes a 201 amino acid protein that contains nuclear factor-binding motifs known to regulate transcription in smooth muscle. During embryogenesis, transgelin is expressed in smooth, cardiac and skeletal muscle, but is restricted during late fetal development and adulthood to all vascular and visceral smooth muscle cells and low levels of expression in heart. Transgelin is down regulated in several transformed cell lines, indicating that a reduction of transgelin expression may be an early indicator of the onset of transformation. Transgelin also binds Actin, causing Actin fibers to gel within minutes of binding. Binding of transgelin to Actin occurs at a ratio of 1:6 Actin monomers.
Target Species Human, Mouse, Pig, Primate, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.