Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase-2, FN binding domain Mouse anti-Human, Unconjugated, Clone: 4G3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3839I100UG
Description
Anti-Transglutaminase-2, FN binding domain, clone 4G3, Cat. No. MAB3839-I, is a mouse monoclonal antibody that detects Protein-glutamine gamma-glutamyltransferase 2 and has been tested for use in Flow Cytometry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Specifications
Transglutaminase-2, FN binding domain | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG1 in PBS without azide. | |
TGM2 | |
A full-length purified tissue transglutaminase (TG-2) from human erythrocytes. | |
Protein G purified | |
RUO | |
Primary | |
Clone 4G3 specifically detects human Transglutaminase-2. | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Purified |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
4G3 | |
Unconjugated | |
Protein-glutamine gamma-glutamyltransferase 2;EC: 2.3.2.13;Tissue transglutaminase;Transglutaminase C;TG(C);TGC;TGase C;Transglutaminase H;TGase H;TGase-2 | |
Mouse | |
∽80 kDa observed; 77.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μg | |
Signaling | |
NP_001310245 | |
Human | |
Antibodies | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction