Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase-2, FN binding domain Mouse anti-Human, Unconjugated, Clone: 4G3, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | Transglutaminase-2, FN binding domain |
---|---|
Clone | 4G3 |
Concentration | Please refer to lot specific datasheet. |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3839-I-25
|
MilliporeSigma
MAB3839I25UG |
25 μg |
Each for $192.35
|
|
|||||
MAB3839-I-100
|
MilliporeSigma
MAB3839I100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-Transglutaminase-2, FN binding domain, clone 4G3, Cat. No. MAB3839-I, is a mouse monoclonal antibody that detects Protein-glutamine gamma-glutamyltransferase 2 and has been tested for use in Flow Cytometry, Immunofluorescence, Immunoprecipitation, and Western Blotting.Specifications
Transglutaminase-2, FN binding domain | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Protein-glutamine gamma-glutamyltransferase 2;EC: 2.3.2.13;Tissue transglutaminase;Transglutaminase C;TG(C);TGC;TGase C;Transglutaminase H;TGase H;TGase-2 | |
TGM2 | |
IgG1 κ | |
Protein G purified | |
Clone 4G3 specifically detects human Transglutaminase-2. | |
∽80 kDa observed; 77.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
4G3 | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Signaling | |
Purified mouse monoclonal antibody IgG1 in PBS without azide. | |
NP_001310245 | |
A full-length purified tissue transglutaminase (TG-2) from human erythrocytes. | |
Primary | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title