Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transglutaminase 2/TGM2 Antibody (CUB7402), Novus Biologicals™
SDP

Catalog No. NBP226458 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226458 0.25 mL
NB019408 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP226458 Supplier Novus Biologicals Supplier No. NBP226458
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transglutaminase 2/TGM2 Monoclonal antibody specifically detects Transglutaminase 2/TGM2 in Human, Guinea Pig samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Transglutaminase 2/TGM2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CUB7402
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:75, Immunohistochemistry-Paraffin 1:50-1:75
Formulation PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
Gene Alias C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2
Gene Symbols TGM2
Host Species Mouse
Immunogen BALB/C mice were injected with purified guinea pig liver TGase II.
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immune System Diseases, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7052
Target Species Human, Guinea Pig, Rabbit
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.