Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transglutaminase 5 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342119 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342119 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342119 Supplier Novus Biologicals Supplier No. NBP281857MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transglutaminase 5 Polyclonal antibody specifically detects Transglutaminase 5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Transglutaminase 5
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.3.2.13, protein-glutamine gamma-glutamyltransferase 5, TG(X), TGase X, TGase-5, TGASE5, TGASEX, TGM6, TGMX, TGXMGC141907, transglutaminase 5, transglutaminase V, Transglutaminase X, transglutaminase-5
Host Species Rabbit
Immunogen Transglutaminase 5 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human Transglutaminase 5. The immunogen is located within the last 50 amino acids of Transglutaminase 5. Amino Acid Squence: NMRSNKFKDIKGYRNVYVD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9333
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.