Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transglutaminase 7/TGM7 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB164926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164926 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164926 Supplier Novus Biologicals Supplier No. NBP281763JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transglutaminase 7/TGM7 Polyclonal antibody specifically detects Transglutaminase 7/TGM7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Transglutaminase 7/TGM7
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias protein-glutamine gamma-glutamyltransferase Z, TG(Z), TGase Z, TGase-7, TGMZEC 2.3.2.13, TGZ, transglutaminase 7, Transglutaminase Z, transglutaminase-7
Host Species Rabbit
Immunogen TGM7 antibody was raised against a 13 amino acid synthetic peptide near the center of human TGM7. The immunogen is located within amino acids 460 - 510 of TGM7. Amino Acid Squence: LESGGLRDQPAQL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 116179
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.