Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transketolase Antibody (OTI5H3), PE, Novus Biologicals™
SDP

Catalog No. NBP274585PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274585PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274585PE Supplier Novus Biologicals Supplier No. NBP274585PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transketolase Monoclonal specifically detects Transketolase in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Transketolase
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5H3
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.2.1.1, FLJ34765, TK, TKT1, transketolase
Gene Symbols TKT
Host Species Mouse
Immunogen Full length recombinant protein of human Transketolase (NP_001055) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7086.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.