Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Globe Scientific Translucent Snap Caps
Click to view available options
Color:
Clear
Gray
Lavender
Red
Description
- These caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures
- They are available in bright translucent colors for sample identification purposes
- The caps feature a single thumb tab for easy removal and close nesting in tube racks
- Features :
- Economical way to recap glass tubes, regardless of manufacturer
- Prevents spills and evaporation by employing a liquid tight seal
- Available in seven translucent colors that correspond to Vacutainer™ cap colors: Natural, Blue, Green, Lavender, Red, Gray, and Yellow
- Caps can be written on with indelible ink for easy sample identification
Specifications
Specifications
| Product Type | Snap Cap |
| Autoclavable | Not Autoclavable |
| Material | LDPE |
| Color | Gray |
| Sterility | Non-sterile |
| For Use With (Equipment) | For Vacuum and Test Tubes |
| Disposable | Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction