Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transportin 1 Antibody (D45) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB014064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014064 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014064 Supplier Novus Biologicals Supplier No. NBP280991
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transportin 1 Monoclonal specifically detects Transportin 1 in Human, Mouse, Bovine, Canine, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray, Flow (Intracellular), CyTOF-ready.

Specifications

Antigen Transportin 1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D45
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000, Flow Cytometry, Immunohistochemistry : 1:200-1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 9298975), Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000. Use reported in scientific literature (PMID 24647604), Microarray, Flow (Intracellular), CyTOF-ready
Formulation PBS
Gene Alias importin beta 2, Importin beta-2, IPO2, karyopherin (importin) beta 2, MIP1Karyopherin beta-2, MIPM9 region interaction protein, transportin 1, transportin-1, TRNKPNB2importin 2
Gene Symbols TNPO1
Host Species Mouse
Immunogen Human recombinant His-tagged Transportin 1. [UniProt# Q92973]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, DNA replication Transcription Translation and Splicing, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3842
Test Specificity Mouse reactivity reported in scientific literature (PMID: 15738418).
Target Species Human, Mouse, Bovine, Canine, Monkey, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.