Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transportin 1 Antibody (D45), PE, Novus Biologicals™
SDP

Catalog No. NB6001397PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001397PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001397PE Supplier Novus Biologicals Supplier No. NB6001397PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transportin 1 Monoclonal specifically detects Transportin 1 in Human, Mouse, Bovine, Canine, Monkey, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen Transportin 1
Applications Flow Cytometry
Classification Monoclonal
Clone D45
Conjugate PE
Dilution Flow Cytometry, Flow (Intracellular)
Formulation PBS with 0.05% Sodium Azide
Gene Alias importin beta 2, Importin beta-2, IPO2, karyopherin (importin) beta 2, MIP1Karyopherin beta-2, MIPM9 region interaction protein, transportin 1, transportin-1, TRNKPNB2importin 2
Gene Symbols TNPO1
Host Species Mouse
Immunogen Human recombinant His-tagged Transportin 1. [UniProt# Q92973]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 3842
Target Species Human, Bovine, Canine, Monkey, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.