Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transthyretin/Prealbumin Antibody (TTR/4292), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB384581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384581 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384581 Supplier Novus Biologicals Supplier No. NBP308765UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transthyretin/Prealbumin Monoclonal antibody specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Transthyretin/Prealbumin
Applications Western Blot, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone TTR/4292
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin
Host Species Mouse
Immunogen Recombinant human full-length protein (Uniprot: P02766)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7276
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.