Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAP1 Antibody (3H4-2H6), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP259343UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259343UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259343UV Supplier Novus Biologicals Supplier No. NBP259343UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAP1 Monoclonal specifically detects TRAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TRAP1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3H4-2H6
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein
Gene Symbols TRAP1
Host Species Mouse
Immunogen Purified recombinant TRAP1
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10131
Test Specificity Detects 75kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.