Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAP1 Rabbit anti-Human, Mouse, Clone: 1L8C1, Novus Biologicals™
SDP

Catalog No. NB167697 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167697 100 μg
NB167698 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167697 Supplier Novus Biologicals Supplier No. NBP316303100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TRAP1 Monoclonal antibody specifically detects TRAP1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TRAP1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 1L8C1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human TRAP1 (Q12931).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10131
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.