Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAP220/MED1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1002574R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002574R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002574R Supplier Novus Biologicals Supplier No. NB1002574R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRAP220/MED1 Polyclonal specifically detects TRAP220/MED1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen TRAP220/MED1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Activator-recruited cofactor 205 kDa component, ARC205, CRSP1TRIP2, CRSP200p53 regulatory protein RB18A, DRIP230PPAR-binding protein, mediator complex subunit 1DRIP205, mediator of RNA polymerase II transcription subunit 1, PBPPPARGBP, Peroxisome proliferator-activated receptor-binding protein, PPAR binding protein, PPARBPMGC71488, PPARG binding protein, RB18ATR-interacting protein 2, Thyroid hormone receptor-associated protein complex 220 kDa component, thyroid hormone receptor-associated protein complex component TRAP220, thyroid receptor interacting protein 2, Thyroid receptor-interacting protein 2, Trap220, TRAP220TRIP-2, vitamin D receptor-interacting protein 230 kD, Vitamin D receptor-interacting protein complex component DRIP205
Gene Symbols MED1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1525 and the C-terminus (residue 1581) of human Cofactor Required for Sp1 Transcriptional Activation 1using the numbering given in entry NP_004765.2 (GeneID 5469).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5469
Test Specificity CRSP1
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.