Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAPPC1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100988110.1MG
Description
TRAPPC1 Polyclonal specifically detects TRAPPC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TRAPPC1 | |
Polyclonal | |
Unconjugated | |
PBS with No Preservative | |
BET5BET5 homolog, Multiple myeloma protein 2, MUM-2, MUM2melanoma ubiquitous mutated 2, trafficking protein particle complex 1, trafficking protein particle complex subunit 1 | |
Rabbit | |
IgG purified | |
RUO | |
58485 | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
LYOPH | |
Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Q9Y5R8 | |
TRAPPC1 | |
A synthetic peptide from the internal part of human internal part of Trafficking protein particle complex subunit 1 conjugated to an immunogenic carrier protein was used as the immunogen. The immunogen is homologous with the corresponding sequence in mouse, rat and chicken. | |
0.1 mg | |
Primary | |
TPPC1. | |
Rat, Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction