Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAPPC1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | TRAPPC1 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017951
![]() |
Novus Biologicals
NB100988110.1MG |
0.1 mg |
Each for $206.50
|
|
|||||
NB10098811
![]() |
Novus Biologicals
NB10098811 |
0.5 mg |
Each for $499.50
|
|
|||||
Description
TRAPPC1 Polyclonal specifically detects TRAPPC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TRAPPC1 | |
Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
BET5BET5 homolog, Multiple myeloma protein 2, MUM-2, MUM2melanoma ubiquitous mutated 2, trafficking protein particle complex 1, trafficking protein particle complex subunit 1 | |
TRAPPC1 | |
Primary | |
TPPC1. |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Q9Y5R8 | |
58485 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title