Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAPPC1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10098811
Description
TRAPPC1 Polyclonal specifically detects TRAPPC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TRAPPC1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from PBS with No Preservative | |
BET5BET5 homolog, Multiple myeloma protein 2, MUM-2, MUM2melanoma ubiquitous mutated 2, trafficking protein particle complex 1, trafficking protein particle complex subunit 1 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
58485 | |
Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
LYOPH | |
Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml | |
Q9Y5R8 | |
TRAPPC1 | |
A synthetic peptide from human TRAPPC1 conjugated to an immunogenic carrier protein was used as the antigen. The peptide is homologous with the corresponding sequence in mouse, rat and chicken. | |
0.5 mg | |
Primary | |
TPPC1. | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction