Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC5 Antibody, Novus Biologicals™
SDP

Catalog No. NB173759 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173759 Supplier Novus Biologicals Supplier No. NBP318481
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRAPPC5 Polyclonal antibody specifically detects TRAPPC5 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TRAPPC5
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 20-40 ug/mg lysate
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias MGC52424, trafficking protein particle complex 5, trafficking protein particle complex subunit 5, TRS31
Host Species Rabbit
Immunogen Recognizes region between residues Between between 138 and 188..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126003
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.