Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trappin-2/Elafin/Skalp Antibody (101), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB334687 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334687 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334687 Supplier Novus Biologicals Supplier No. NBP290071JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Trappin-2/Elafin/Skalp Monoclonal antibody specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for ELISA

Specifications

Antigen Trappin-2/Elafin/Skalp
Applications ELISA
Classification Monoclonal
Clone 101
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias cementoin, Elastase-specific inhibitor, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human Trappin-2/Elafin/Skalp (Accession#: NP_002629.1, Met1-Gln117)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5266
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.