Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | TRAT1 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8620
![]()
|
Proteintech Group Inc
132791AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TRIM (T cell receptor-interacting molecule), also known as TRAT1 (T cell receptor associated transmembrane adaptor 1) is a 30 kDa protein expressed by T cells as a cystein-linked homodimer. It associates with TCR-CD3-zeta-chain complex and becomes phosphorylated by Src-family kinases. TRIM is potentially involved in negative regulation of TCR-mediated signaling, but its role remains unclear.Specifications
| TRAT1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q3UU67, Q6PIZ9 | |
| 498075, 50852, 77647 | |
| TRAT1 Fusion Protein Ag4023 | |
| Primary | |
| -20°C | |
| TRAT1 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HSPC062, pp29/30, TCRIM, TRAT1, TRIM | |
| TRAT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title