Learn More
Eisco™ Travertine Sedimentary Rock Specimen
SCI_ED
Catalog No. p-7203018
Click to view available options
Quantity:
1/Ea.
12/Pk.
Travertine is a carbonate rock formed from gushing hot springs. The airy, porous material indicates an abundance of air bubbles moving through the rock as it solidified. Travertine is unique in that it forms as a result of rapidly dropping pH levels.
  • Correlates well to Next Generation Science Standards Evidence
  • Ideal for teaching students about rounding and sorting, as well as for identification & classification exercises
  • Useful in lessons on sedimentary petrology and geothermal systems
Specifications
Product Type Sedimentary Rock
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.