Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRBC1 Antibody (JOVI.1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB154883 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154883 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB154883 Supplier Novus Biologicals Supplier No. NBP305752

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRBC1 Monoclonal antibody specifically detects TRBC1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)

Specifications

Antigen TRBC1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone JOVI.1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 2-6 ug/mL, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias BV05S1J2.2, TCRBC1, TRBC1 T cell receptor beta constant 1
Host Species Mouse
Immunogen Thymus, spleen, and mesenteric lymph nodes isolated from a mouse transgenic for human TCR Vbeta3Cbeta1
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28639
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG2a Kappa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.