Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRBP Antibody (PCRP-TARBP2-1E5), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB387731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387731 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387731 Supplier Novus Biologicals Supplier No. NBP308235DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRBP Monoclonal antibody specifically detects TRBP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen TRBP
Applications Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-TARBP2-1E5
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias LOQS, RISC-loading complex subunit TARBP2, TAR (HIV) RNA binding protein 2, TAR (HIV) RNA-binding protein 2, TAR (HIV) RNA-binding protein TRBP1, TAR (HIV-1) RNA binding protein 2, TAR RNA binding protein 2, TAR RNA-binding protein 2, trans-activation responsive RNA-binding protein, Trans-activation-responsive RNA-binding protein, TRBP, TRBP1, TRBP2
Host Species Mouse
Immunogen Recombinant full-length human TRBP protein (Uniprot: Q15633)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6895
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.