Learn More
Johannes Lieder Trees and Shrubs (Hazel, Chestnut, Willow, Beech, Oak)
Shop All Johannes Lieder GmbH & Co. KG Products
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 6230 Trees and Shrubs
- 12 Microscope Slides
- Hazel (Corylus avellana), female flower l.s.
- Hazel, male flower l.s.
- Hazel, mature pollen grains w.m.
- Hazel, young fruit (nut) l.s.
- Willow (Salix alba), young aggregate fruit l.s.
- Horse chestnut (Aesculus hippocastanum), petiole t.s.
- Horse chestnut, leaf bud t.s., shows leaf origin
- Horse chestnut, flower bud l.s.
- Horse chestnut, young fruit t.s.
- Beech (Fagus silvatica), sun and shadow leaves t.s.
- Beech, wood sections: transverse, tangential, radial
- Oak (Quercus robur), wood sections: transverse, tangential, radial
Specifications
Specifications
Classification | Botany |
Material | Elastin Stained Glass |
Mounting Type | Cross and Longitudinal Section |
Type | Microscope Slides |
Slide Type | Prepared Slide |