Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREK 1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB11041535C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11041535C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11041535C Supplier Novus Biologicals Supplier No. NB11041535C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TREK 1 Polyclonal specifically detects TREK 1 in Human, Mouse, Bovine samples. It is validated for Western Blot.

Specifications

Antigen TREK 1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot 2 ug/ml
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O95069
Gene Alias MGC126742, MGC126744, potassium channel, subfamily K, member 2, subfamily K, member 2, tandem-pore-domain potassium channel TREK-1, TREK-1 K(+) channel subunit, TWIK-related potassium channel 1, two-pore potassium channel 1
Gene Symbols KCNK2
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3776
Test Specificity Human, mouse, and cow. 88% sequence identity with rat protein.
Target Species Human, Mouse, Bovine
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.