Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM-2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $440.96
Specifications
Antigen | TREM-2 |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN1471-25UG
|
MilliporeSigma
ABN147125UG |
25 μg |
Each for $192.35
|
|
|||||
ABN1471MI
|
MilliporeSigma
ABN1471 |
100 μg |
Each for $440.96
|
|
|||||
Description
Anti-TREM-2 is a highly specific rabbit polyclonal antibody that targets Triggering receptor expressed on myeloid cells 2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
TREM-2 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of human TREM-2. | |
Affinity Purified | |
This polyclonal antibody antibody detects Triggering receptor expressed on myeloid cells 2 in several regions of human brain. It targets an epitope within 13 amino acids from the C-terminal region. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Neuroscience | |
TREM2 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title