Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM1 Antibody (174031) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB400700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400700 Supplier Novus Biologicals Supplier No. FAB1187Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREM1 Monoclonal antibody specifically detects TREM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF

Specifications

Antigen TREM1
Applications Western Blot, Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 174031
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TREM-1, Ala21-Ser202, Accession # Q9JKE2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54210
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.